4-Bromo-2-nitroanisole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Bromo-1-methoxy-2-nitrobenzene
B4010-5G
B4010-25G
TCIAB4010-5GEA
70
EUR
TCIAB4010-5G
TCIAB4010-25G
4-Bromo-2-nitroanisole ≥98.0% (by GC)
4-Bromo-2-nitroanisole
Formula:
C₇H₆BrNO₃ Peso molecolare: 232,03 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00055529 CAS Number: 33696-00-3 EINECS: 251-642-5 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...