2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)
Fornitore: TCI EUROPE
B3997-1G
B3997-5G
TCIAB3997-1GEA
32
EUR
TCIAB3997-1G
TCIAB3997-5G
2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)
2-Bromo-5-hydroxybenzaldehyde
Formula:
C₇H₅BrO₂ Peso molecolare: 201,02 g/mol |
MDL Number:
MFCD03033065 CAS Number: 2973-80-0 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...