4-(Bromomethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4-(Bromomethyl)-4'-methyl-2,2'-bipyridine
Formula:
C₁₂H₁₁BrN₂ |
MDL Number:
MFCD08275265 CAS Number: 81998-05-2 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 97.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...