2,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,5-Bis(trifluoromethyl)benzoylchloride, 2,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride
B3980-1G
B3980-5G
TCIAB3980-1GEA
39
EUR
TCIAB3980-1G
TCIAB3980-5G
2,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)
2,5-Bis(trifluoromethyl)benzoyl chloride
Formula:
C₉H₃ClF₆O Peso molecolare: 276,57 g/mol Punto di ebollizione: 72°C Flash Pt: 74°C |
MDL Number:
MFCD00074997 CAS Number: 393-82-8 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...