Stampa…

1-(4-Bromophenyl)naphthalene ≥98.0% (by GC)

Fornitore: TCI EUROPE

B3978-1G B3978-5G
TCIAB3978-1GEA 17 EUR
TCIAB3978-1G TCIAB3978-5G
1-(4-Bromophenyl)naphthalene ≥98.0% (by GC)
1-(4-Bromophenyl)naphthalene
Formula: C₁₆H₁₁Br
Peso molecolare: 283,17 g/mol
MDL Number: MFCD07369916
CAS Number: 204530-94-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR