Stampa…

9-Bromo-10-phenylanthracene ≥98.0% (by GC)

Fornitore: TCI

B3977-1G B3977-5G
TCIAB3977-1GEA 26 EUR
TCIAB3977-1G TCIAB3977-5G
9-Bromo-10-phenylanthracene ≥98.0% (by GC)
9-Bromo-10-phenylanthracene
Formula: C₂₀H₁₃Br
MDL Number: MFCD00230983
CAS Number: 23674-20-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR