Stampa…

Boc-glycine methyl ester ≥98.0% (by total nitrogen basis)

Fornitore: TCI

Synonyms: N-Boc-glycine Methyl Ester, Boc-Gly-OMe

B3975-5G B3975-25G
TCIAB3975-5GEA 25 EUR
TCIAB3975-5G TCIAB3975-25G
Boc-glycine methyl ester ≥98.0% (by total nitrogen basis)
Boc-glycine methyl ester
Formula: C₈H₁₅NO₄
Peso molecolare: 189.2 g/mol
Flash Pt: 109°C
MDL Number: MFCD00038267
CAS Number: 31954-27-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR