tert-Butyl pyrrolidine-1-carboxylate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
tert-butyl pyrrolidine-1-carboxylate, pyrrolidine-1-carboxylic acid tert-butyl ester, N-BOC-Pyrrolidine
Formula:
C₉H₁₇NO₂ Punto di ebollizione: 75°C Flash Pt: 86°C |
MDL Number:
MFCD00216581 CAS Number: 86953-79-9 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...