5-Bromo-2-methoxybenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
B3965-1G
B3965-5G
TCIAB3965-1GEA
30
EUR
TCIAB3965-1G
TCIAB3965-5G
5-Bromo-2-methoxybenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
5-Bromo-2-methoxybenzenesulphonyl chloride
Formula:
C₇H₆BrClO₃S Peso molecolare: 285,55 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00051768 CAS Number: 23095-05-8 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...