1-Boc-4-cyanopiperidine ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₁H₁₈N₂O₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01861223 CAS Number: 91419-52-2 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...