Stampa…

9-Bromotriptycene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene

B3955-1G B3955-5G
TCIAB3955-1GEA 100 EUR
TCIAB3955-1G TCIAB3955-5G
9-Bromotriptycene ≥98.0% (by GC)
9-Bromotriptycene
Formula: C₂₀H₁₃Br
Peso molecolare: 333,23 g/mol
MDL Number: MFCD01099022
CAS Number: 15364-55-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 252.0 to 256.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR