Stampa…

4-Bromo-2-chlorobenzaldehyde ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloro-4-bromobenzaldehyde

B3948-1G B3948-5G
TCIAB3948-1GEA 21 EUR
TCIAB3948-1G TCIAB3948-5G
4-Bromo-2-chlorobenzaldehyde ≥97.0% (by GC)
4-Bromo-2-chlorobenzaldehyde
Formula: C₇H₄BrClO
Peso molecolare: 219,46 g/mol
MDL Number: MFCD08741413
CAS Number: 158435-41-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR