2-Bromophenethylamine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-(2-Bromophenyl)ethylamine
B3938-5G
B3938-25G
TCIAB3938-5GEA
69
EUR
TCIAB3938-5G
TCIAB3938-25G
2-Bromophenethylamine ≥98.0% (by titrimetric analysis)
2-Bromophenethylamine
Formula:
C₈H₁₀BrN Peso molecolare: 200,08 g/mol Punto di ebollizione: 85°C |
MDL Number:
MFCD01529874 CAS Number: 65185-58-2 UN: 2735 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...