1,2-Benzofluorene ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
11H-Benzo[a]fluorene
B3931-200MG
B3931-1G
TCIAB3931-200MGEA
123
EUR
TCIAB3931-200MG
TCIAB3931-1G
1,2-Benzofluorene ≥95.0% (by GC)
1,2-Benzofluorene
Formula:
C₁₇H₁₂ Peso molecolare: 216,28 g/mol Punto di ebollizione: 407°C |
MDL Number:
MFCD00039578 CAS Number: 238-84-6 EINECS: 205-944-9 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 181.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...