1-Bromo-2,5-difluoro-4-nitrobenzene ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₂BrF₂NO₂ |
MDL Number:
MFCD00042184 CAS Number: 167415-27-2 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...