Stampa…

3-Bromo-9-phenyl-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Bromo-9-phenylcarbazole

B3908-1G B3908-5G
TCIAB3908-1GEA 16 EUR
TCIAB3908-1G TCIAB3908-5G
3-Bromo-9-phenyl-9H-carbazole ≥98.0% (by GC)
3-Bromo-9-phenyl-9H-carbazole
Formula: C18H12BrN
Peso molecolare: 322,20 g/mol
MDL Number: MFCD11977305
CAS Number: 1153-85-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR