Stampa…

4-Benzyl-N-carbobenzoxy-L-aspartate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 4-Benzyl N-Cbz-L-aspartate, N-Carbobenzoxy-L-aspartic Acid 4-Benzyl Ester, N-Cbz-L-aspartic Acid 4-Benzyl Ester, Z-Asp(OBzl)-OH, Cbz-Asp(OBzl)-OH

B3903-25G B3903-5G
TCIAB3903-25GEA 94 EUR
TCIAB3903-25G TCIAB3903-5G
4-Benzyl-N-carbobenzoxy-L-aspartate ≥98.0% (by HPLC, titration analysis)
4-Benzyl-N-carbobenzoxy-L-aspartate
Formula: C₁₉H₁₉NO₆
Peso molecolare: 357,36 g/mol
MDL Number: MFCD00037820
CAS Number: 3479-47-8
EINECS: 222-456-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C
Specific rotation [a]20/D +11.0 to +14.0 deg (C=1、AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR