4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Fmoc-Asp(OBzl)-OH, 4-Benzyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester
B3887-25G
B3887-5G
TCIAB3887-25GEA
228
EUR
TCIAB3887-25G
TCIAB3887-5G
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate ≥98.0% (by HPLC, titration analysis)
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
Formula:
C₂₆H₂₃NO₆ Peso molecolare: 445,47 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00065630 CAS Number: 86060-84-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Specific rotation [a]20/D | -18.0 to -22.0 deg (C=1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...