Stampa…

2-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B3879-1G B3879-5G
TCIAB3879-1GEA 24 EUR
TCIAB3879-1G TCIAB3879-5G
2-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)
2-Bromo-p-toluic acid
Formula: C₈H₇BrO₂
Peso molecolare: 215,05 g/mol
MDL Number: MFCD00045797
CAS Number: 7697-27-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR