Stampa…

tert-Butyl-4-formylpiperidine-1-carboxylate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: tert-Butyl 4-formylpiperidine-1-carboxylate, N-Boc-4-Formylpiperidine, 1-BOC-4-Piperidinecarboxaldehyde, 1-Boc-4-formylpiperidine, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehyde, 1-(tert-Butoxycarbonyl)-4-formylpiperidine, 1-Boc-4-piperidinecarboxaldehyde, Tert-butyl 4-formylpiperidine-1-carboxylate

B3873-1G B3873-5G
TCIAB3873-1GEA 30 EUR
TCIAB3873-1G TCIAB3873-5G
tert-Butyl-4-formylpiperidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-4-formylpiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
Peso molecolare: 213,28 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Melting point 32.0 to 37.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR