Stampa…

5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 5-Carboxybenzotriazole

B3869-5G B3869-25G
TCIAB3869-5GEA 41 EUR
TCIAB3869-5G TCIAB3869-25G
5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)
5-Benzotriazolecarboxylic acid
Formula: C₇H₅N₃O₂
Peso molecolare: 163,14 g/mol
MDL Number: MFCD00012318
CAS Number: 23814-12-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR