Stampa…

Bis(aminomethyl)norbornane (mixture of isomers) ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Bicyclo[2.2.1]heptanedimethanamine, Bis(aminomethyl)bicyclo[2.2.1]heptane

B3852-25G B3852-500G
TCIAB3852-25GEA 16 EUR
TCIAB3852-25G TCIAB3852-500G
Bis(aminomethyl)norbornane (mixture of isomers) ≥98.0% (by titrimetric analysis)
Bis(aminomethyl)norbornane (mixture of isomers)
Formula: C₉H₁₈N₂
Peso molecolare: 154,26 g/mol
CAS Number: 56602-77-8
EINECS: 260-280-7
UN: 2735
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR