Stampa…

5-Bromo-o-anisidine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

B3828-1G B3828-5G
TCIAB3828-1GEA 28 EUR
TCIAB3828-1G TCIAB3828-5G
5-Bromo-o-anisidine ≥97.0% (by titrimetric analysis)
5-Bromo-o-anisidine
Formula: C₇H₈BrNO
Peso molecolare: 202,05 g/mol
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR