Stampa…
1-Boc-3-oxopiperazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine
B3804-1G B3804-5G
TCIAB3804-1GEA 23 EUR
TCIAB3804-1G TCIAB3804-5G
1-Boc-3-oxopiperazine ≥98.0% (by GC)
1-Boc-3-oxopiperazine
Formula: C₉H₁₆N₂O₃
Peso molecolare: 200,238 g/mol
MDL Number: MFCD02181069
CAS Number: 76003-29-7
Visualizza i codici
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 164.0 deg-C