Stampa…

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,4-Benzodioxane-6-carboxylic acid

B3764-5G B3764-25G
TCIAB3764-5GEA 49 EUR
TCIAB3764-5G TCIAB3764-25G
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid
Formula: C₉H₈O₄
Peso molecolare: 180,16 g/mol
MDL Number: MFCD00463509
CAS Number: 4442-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc811abf8161a48d967af16a9752088616969f0a&vid=fc811abf8161a48d967af16a9752088616969f0a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR