Stampa…

2-Bromobenzyl mercaptan ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Bromobenzylmercaptan, o-Bromotoluene-α-thiol

B3747-5G B3747-25G
TCIAB3747-5GEA 131 EUR
TCIAB3747-5G TCIAB3747-25G
2-Bromobenzyl mercaptan ≥98.0% (by GC)
2-Bromobenzyl mercaptan
Formula: C₇H₇BrS
Peso molecolare: 203,1 g/mol
Punto di ebollizione: 82°C
MDL Number: MFCD04038925
CAS Number: 143888-85-1
UN: 2810
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR