6-Bromo-4,4'-dimethyl-2,2'-bipyridyl ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromo-4,4'-dimethyl-2,2'-bipyridine
B3732-1G
B3732-5G
TCIAB3732-1GEA
185
EUR
TCIAB3732-1G
TCIAB3732-5G
6-Bromo-4,4'-dimethyl-2,2'-bipyridyl ≥97.0% (by GC)
6-Bromo-4,4'-dimethyl-2,2'-bipyridyl
Formula:
C₁₂H₁₁BrN₂ Peso molecolare: 263,14 g/mol |
MDL Number:
MFCD11042495 CAS Number: 850413-36-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...