5-Bromonicotinonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine
B3713-1G
B3713-5G
TCIAB3713-1GEA
50
EUR
TCIAB3713-1G
TCIAB3713-5G
5-Bromonicotinonitrile ≥98.0% (by GC)
5-Bromonicotinonitrile
Formula:
C₆H₃BrN₂ Peso molecolare: 183,01 g/mol |
MDL Number:
MFCD00174363 CAS Number: 35590-37-5 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...