Stampa…

(S)-(-)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (S)-1-(4-Bromophenyl)ethanamine, (S)-(-)-1-(4-Bromophenyl)ethylamine

B3674-25G B3674-5G
TCIAB3674-25GEA 365 EUR
TCIAB3674-25G TCIAB3674-5G
(S)-(-)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-4-Bromo-α-methylbenzylamine
Formula: C₈H₁₀BrN
Peso molecolare: 200,08 g/mol
MDL Number: MFCD00066026
CAS Number: 27298-97-1
UN: 2735
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -21.0 deg (C=3、MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR