Stampa…

Boc-N-methyl-D-alanine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

B3651-25G B3651-5G
TCIAB3651-25GEA 55 EUR
TCIAB3651-25G TCIAB3651-5G
Boc-N-methyl-D-alanine ≥98.0% (by GC, titration analysis)
Boc-N-methyl-D-alanine
Formula: C₉H₁₇NO₄
Peso molecolare: 203,2 g/mol
MDL Number: MFCD00037242
CAS Number: 16948-16-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 89.0 to 94.0 deg-C
Specific rotation [a]20/D -29.0 to -32.0 deg (C=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR