Stampa…

9,9-Bis(4-bromophenyl)fluorene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 9,9-Bis(4-bromophenyl)-9H-fluorene

B3645-1G B3645-5G
TCIAB3645-1GEA 146 EUR
TCIAB3645-1G TCIAB3645-5G
9,9-Bis(4-bromophenyl)fluorene ≥98.0% (by GC)
9,9-Bis(4-bromophenyl)fluorene
Formula: C₂₅H₁₆Br₂
Peso molecolare: 476,21 g/mol
MDL Number: MFCD19707068
CAS Number: 128406-10-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR