Stampa…

1-(4-Bromophenyl)-1,2,2-triphenylethylene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Bromo-4-(1,2,2-triphenylethenyl)benzene

B3634-1G B3634-5G
TCIAB3634-1GEA 60 EUR
TCIAB3634-1G TCIAB3634-5G
1-(4-Bromophenyl)-1,2,2-triphenylethylene ≥98.0% (by GC)
1-(4-Bromophenyl)-1,2,2-triphenylethylene
Formula: C₂₆H₁₉Br
Peso molecolare: 411,34 g/mol
MDL Number: MFCD20257904
CAS Number: 34699-28-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR