N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine ≥97.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
B3631-1G
B3631-5G
TCIAB3631-1GEA
212
EUR
TCIAB3631-1G
TCIAB3631-5G
N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine ≥97.0% (by HPLC, total nitrogen)
N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine
Formula:
C₂₈H₄₃N₃O₇Si₂ Punto di fusione: 101°C |
CAS Number:
69304-43-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...