tert-Butyl-N-allylcarbamate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate
B3618-25G
B3618-5G
TCIAB3618-25GEA
117
EUR
TCIAB3618-25G
TCIAB3618-5G
tert-Butyl-N-allylcarbamate ≥98.0% (by GC)
tert-Butyl-N-allylcarbamate
Formula:
C₈H₁₅NO₂ Peso molecolare: 157,21 g/mol |
MDL Number:
MFCD00191870 CAS Number: 78888-18-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...