Stampa…

Benazepril hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-((S)-3-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid hydrochloride, (3S)-3-[(1S)-1-Ethoxycarbonyl-3-phenylpropyl]amino-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid Hydrochloride

B3611-1G B3611-5G
TCIAB3611-1GEA 90 EUR
TCIAB3611-1G TCIAB3611-5G
Benazepril hydrochloride ≥98.0% (by HPLC, titration analysis)
Benazepril hydrochloride
Formula: C₂₄H₂₈N₂O₅
Peso molecolare: 460,96 g/mol
Punto di fusione: 183°C
MDL Number: MFCD00895734
CAS Number: 86541-74-4
Merck Index: 14,01031

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -134.0 to -140.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR