4-Bromo-2,2-diphenylbutyric acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B3597-25G
B3597-5G
TCIAB3597-25GEA
161
EUR
TCIAB3597-25G
TCIAB3597-5G
4-Bromo-2,2-diphenylbutyric acid ≥98.0% (by titrimetric analysis)
4-Bromo-2,2-diphenylbutyric acid
Formula:
C₁₆H₁₅BrO₂ Peso molecolare: 319,20 g/mol |
MDL Number:
MFCD01321300 CAS Number: 37742-98-6 EINECS: 253-648-3 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...