tert-Butyl phenyl carbonate ≥96.0% (by HPLC)
Fornitore: TCI
Synonyms:
Carbonic Acid tert-Butyl Phenyl Ester
Formula:
C₁₁H₁₄O₃ Punto di ebollizione: 78°C Flash Pt: 101°C |
MDL Number:
MFCD00008804 CAS Number: 6627-89-0 EINECS: 229-601-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (HPLC) | min. 96.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...