Stampa…

5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromo-7-azaindole

B3588-1G B3588-5G
TCIAB3588-1GEA 30 EUR
TCIAB3588-1G TCIAB3588-5G
5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅BrN₂
Peso molecolare: 197,04 g/mol
MDL Number: MFCD06659677
CAS Number: 183208-35-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4c95a0037bbb7385a849583e769ff1b9d68be83&vid=f4c95a0037bbb7385a849583e769ff1b9d68be83&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR