1,3-Bis(trimethylsilyloxy)propane ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉H₂₄O₂Si₂ Punto di ebollizione: 77°C |
MDL Number:
MFCD00161340 CAS Number: 17887-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...