9-(4-Bromophenyl)carbazole ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
B3554-1G
B3554-5G
TCIAB3554-1GEA
16
EUR
TCIAB3554-1G
TCIAB3554-5G
9-(4-Bromophenyl)carbazole ≥98.0% (by GC)
9-(4-Bromophenyl)carbazole
Formula:
C₁₈H₁₂BrN |
MDL Number:
MFCD11617969 CAS Number: 57102-42-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...