Stampa…

2,2'-Bipyridine-6,6'-dicarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2-Bipyridine-6,6-dicarboxylic Acid

B3533-1G B3533-5G
TCIAB3533-1GEA 148 EUR
TCIAB3533-1G TCIAB3533-5G
2,2'-Bipyridine-6,6'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
2,2'-Bipyridine-6,6'-dicarboxylic acid
Formula: C₁₂H₈N₂O₄
Peso molecolare: 244,21 g/mol
Punto di fusione: 288°C
MDL Number: MFCD11042476
CAS Number: 4479-74-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR