Stampa…

2,6-Bis(2-benzimidazolyl)pyridine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Bis(benzimidazol-2-yl)pyridine, 2,2'-(2,6-Pyridinediyl)bis(benzimidazole)

B3510-1G B3510-5G
TCIAB3510-1GEA 114 EUR
TCIAB3510-1G TCIAB3510-5G
2,6-Bis(2-benzimidazolyl)pyridine ≥98.0% (by HPLC)
2,6-Bis(2-benzimidazolyl)pyridine
Formula: C₁₉H₁₃N₅
Peso molecolare: 311,35 g/mol
Punto di fusione: 353°C
MDL Number: MFCD00134500
CAS Number: 28020-73-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR