Stampa…

2,2'-Bicinchoninic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: [2,2'-Biquinoline]-4,4'-dicarboxylic acid, 2,2'-Biquinoline-4,4'-dicarboxylic Acid, 2,2′-Bicinchoninic acid

B3509-1G B3509-5G
TCIAB3509-1GEA 40 EUR
TCIAB3509-1G TCIAB3509-5G
2,2'-Bicinchoninic acid ≥98.0% (by HPLC)
2,2'-Bicinchoninic acid
Formula: C₂₀H₁₂N₂O₄
Peso molecolare: 344,33 g/mol
Punto di fusione: 365°C
MDL Number: MFCD00068342
CAS Number: 1245-13-2
EINECS: 214-990-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR