2-(Bromoacetyl)-6-methoxynaphthalene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Bromo-6'-methoxy-2'-acetonaphthone
B3496-1G
B3496-5G
TCIAB3496-1GEA
59
EUR
TCIAB3496-1G
TCIAB3496-5G
2-(Bromoacetyl)-6-methoxynaphthalene ≥98.0% (by GC)
2-(Bromoacetyl)-6-methoxynaphthalene
Formula:
C₁₃H₁₁BrO₂ Peso molecolare: 279,13 g/mol |
MDL Number:
MFCD03964189 CAS Number: 10262-65-4 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...