Stampa…

(5-Bromo-3-pyridinyl)methanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3-Bromo-5-(hydroxymethyl)pyridine, 5-Bromo-3-pyridinemethanol

B3493-1G B3493-5G
TCIAB3493-1GEA 78 EUR
TCIAB3493-1G TCIAB3493-5G
(5-Bromo-3-pyridinyl)methanol ≥98.0% (by GC)
(5-Bromo-3-pyridinyl)methanol
Formula: C₆H₆BrNO
Peso molecolare: 188,0200 g/mol
MDL Number: MFCD03265757
CAS Number: 37669-64-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR