Stampa…

(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine

B3485-1G B3485-5G
TCIAB3485-1GEA 23 EUR
TCIAB3485-1G TCIAB3485-5G
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Formula: C₁₈H₂₇NO₅
Peso molecolare: 337,4 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00065598
CAS Number: 47375-34-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C
Specific rotation [a]20/D -14.0 to -18.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR