Stampa…

N,N'-Bis(dimethylaminomethylene)hydrazine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B3480-5G B3480-25G
TCIAB3480-5GEA 119 EUR
TCIAB3480-5G TCIAB3480-25G
N,N'-Bis(dimethylaminomethylene)hydrazine ≥98.0% (by GC, titration analysis)
N,N'-Bis(dimethylaminomethylene)hydrazine
Formula: C₆H₁₄N₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00957712
CAS Number: 16114-05-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR