4-Bromo-N-methylphthalimide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B3443-25G
B3443-5G
TCIAB3443-25GEA
408
EUR
TCIAB3443-25G
TCIAB3443-5G
4-Bromo-N-methylphthalimide ≥98.0% (by GC)
4-Bromo-N-methylphthalimide
Formula:
C₉H₆BrNO₂ |
CAS Number:
90224-73-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...