2,5-Bis(4-biphenylyl)thiophene
Fornitore: TCI EUROPE
Synonyms:
BP1T
B3441-1G
B3441-5G
TCIAB3441-1GEA
101
EUR
TCIAB3441-1G
TCIAB3441-5G
2,5-Bis(4-biphenylyl)thiophene
2,5-Bis(4-biphenylyl)thiophene
Formula:
C₂₈H₂₀S Peso molecolare: 388,53 g/mol Punto di fusione: 337°C |
CAS Number:
56316-86-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Elemental analysis (Sulfuride) | 7.90 to 8.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...