Stampa…

Bis(2-hydroxyethyl)terephthalate ≥85.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: BHET, Terephthalic Acid Bis(2-hydroxyethyl) Ester, Bis(2-hydroxyethyl) terephthalate

B3429-25G B3429-500G
TCIAB3429-25GEA 17 EUR
TCIAB3429-25G TCIAB3429-500G
Bis(2-hydroxyethyl)terephthalate ≥85.0% (by HPLC)
Bis(2-hydroxyethyl)terephthalate
Formula: C₁₂H₁₄O₆
Peso molecolare: 254,24 g/mol
MDL Number: MFCD00081066
CAS Number: 959-26-2
EINECS: 213-497-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 85.0 area%
Melting point 104.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR